Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608399 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSMD3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
AFR
|
African American
|
YRI (Yoruba)
|
|||||||||
EAS - Not Available | Japanese
|
JPT (Japanese)
|
|||||||||
SAS - Not Available | Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
CSMD3 - CUB and Sushi multiple domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052900.2 | 12962 | UTR 3 | NP_443132.3 | |||
NM_198123.1 | 12962 | UTR 3 | NP_937756.1 | |||
NM_198124.1 | 12962 | UTR 3 | NP_937757.1 | |||
XM_011516815.2 | 12962 | UTR 3 | XP_011515117.1 | |||
XM_011516816.2 | 12962 | UTR 3 | XP_011515118.1 | |||
XM_017013008.1 | 12962 | UTR 3 | XP_016868497.1 | |||
XM_017013009.1 | 12962 | UTR 3 | XP_016868498.1 | |||
XM_017013010.1 | 12962 | UTR 3 | XP_016868499.1 | |||
XM_017013011.1 | 12962 | UTR 3 | XP_016868500.1 | |||
XM_017013012.1 | 12962 | UTR 3 | XP_016868501.1 |