Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCGCGGCCCCTACCTCCTAGTT[A/G]CAAGTAGGACCCCCAGTGCCCCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HLF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HLF - HLF, PAR bZIP transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002126.4 | Intron | NP_002117.1 | ||||
XM_005257269.2 | Intron | XP_005257326.1 | ||||
XM_011524705.1 | Intron | XP_011523007.1 | ||||
XM_017024556.1 | Intron | XP_016880045.1 |