Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGATGCCCCTCCAGCCCCAGCCG[C/G]AAAGCCCATACCATTGCCAGTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613293 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3PXD2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH3PXD2B - SH3 and PX domains 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017995.2 | Intron | NP_001017995.1 | ||||
NM_001308175.1 | Intron | NP_001295104.1 | ||||
XM_017009351.1 | Intron | XP_016864840.1 |