Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613423 MIM: 611483 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KCTD16 - potassium channel tetramerization domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020768.3 | Intron | NP_065819.1 | ||||
XM_005268492.2 | Intron | XP_005268549.1 | ||||
XM_005268493.2 | Intron | XP_005268550.1 | ||||
XM_006714789.2 | Intron | XP_006714852.1 | ||||
XM_011537671.2 | Intron | XP_011535973.1 | ||||
XM_011537672.2 | Intron | XP_011535974.1 | ||||
XM_011537673.2 | Intron | XP_011535975.1 |
YIPF5 - Yip1 domain family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |