Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF417 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF417 - zinc finger protein 417 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF418 - zinc finger protein 418 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317027.1 | Intron | NP_001303956.1 | ||||
NM_001317028.1 | Intron | NP_001303957.1 | ||||
NM_001317029.1 | Intron | NP_001303958.1 | ||||
NM_001317030.1 | Intron | NP_001303959.1 | ||||
NM_133460.2 | Intron | NP_597717.1 | ||||
XM_017026304.1 | Intron | XP_016881793.1 | ||||
XM_017026305.1 | Intron | XP_016881794.1 | ||||
XM_017026306.1 | Intron | XP_016881795.1 | ||||
XM_017026307.1 | Intron | XP_016881796.1 | ||||
XM_017026308.1 | Intron | XP_016881797.1 | ||||
XM_017026309.1 | Intron | XP_016881798.1 | ||||
XM_017026310.1 | Intron | XP_016881799.1 | ||||
XM_017026311.1 | Intron | XP_016881800.1 | ||||
XM_017026312.1 | Intron | XP_016881801.1 | ||||
XM_017026313.1 | Intron | XP_016881802.1 | ||||
XM_017026314.1 | Intron | XP_016881803.1 |
Set Membership: |
HapMap |