Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGTTCTTCGGACCTCTGTGCTAC[A/G]GGGACCAGATCAGGGCCAAAGGGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608263 MIM: 610028 | |||||||||||||||||||||||||||||||||||
Literature Links: |
HSPBAP1 PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
HSPBAP1 - HSPB1 associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320728.1 | 783 | Silent Mutation | CCC,CCT | P,P 366 | NP_001307657.1 | |
NM_024610.5 | 783 | Silent Mutation | CCC,CCT | P,P 394 | NP_078886.2 | |
XM_005247762.2 | 783 | Intron | XP_005247819.1 | |||
XM_011513130.2 | 783 | Silent Mutation | CCC,CCT | P,P 357 | XP_011511432.1 | |
XM_011513132.2 | 783 | Silent Mutation | CCC,CCT | P,P 355 | XP_011511434.2 | |
XM_017007179.1 | 783 | Silent Mutation | CCC,CCT | P,P 332 | XP_016862668.1 | |
XM_017007180.1 | 783 | Silent Mutation | CCC,CCT | P,P 159 | XP_016862669.1 |
PARP14 - poly(ADP-ribose) polymerase family member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |