Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607357 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNQ5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KCNQ5 - potassium voltage-gated channel subfamily Q member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160130.1 | Intron | NP_001153602.1 | ||||
NM_001160132.1 | Intron | NP_001153604.1 | ||||
NM_001160133.1 | Intron | NP_001153605.1 | ||||
NM_001160134.1 | Intron | NP_001153606.1 | ||||
NM_019842.3 | Intron | NP_062816.2 | ||||
XM_017011058.1 | Intron | XP_016866547.1 |
KCNQ5-IT1 - KCNQ5 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |