Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602466 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRY2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPRY2 - sprouty RTK signaling antagonist 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318536.1 | 861 | Missense Mutation | ACT,GCT | T,A 106 | NP_001305465.1 | |
NM_001318537.1 | 861 | Missense Mutation | ACT,GCT | T,A 106 | NP_001305466.1 | |
NM_001318538.1 | 861 | Missense Mutation | ACT,GCT | T,A 106 | NP_001305467.1 | |
NM_005842.3 | 861 | Missense Mutation | ACT,GCT | T,A 106 | NP_005833.1 |
Set Membership: |
HapMap |