Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAGGAGAGTGGCGGTAGCATCA[A/C]CAGAAATAGAAAAGTGAAAGATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRMS1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRMS1L - breast cancer metastasis-suppressor 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032352.3 | Intron | NP_115728.2 | ||||
XM_005268128.1 | Intron | XP_005268185.1 | ||||
XM_006720274.2 | Intron | XP_006720337.1 | ||||
XM_006720275.2 | Intron | XP_006720338.1 | ||||
XM_017021703.1 | Intron | XP_016877192.1 | ||||
XM_017021704.1 | Intron | XP_016877193.1 | ||||
XM_017021705.1 | Intron | XP_016877194.1 | ||||
XM_017021706.1 | Intron | XP_016877195.1 |