Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609416 MIM: 609415 MIM: 609417 MIM: 609418 MIM: 609419 MIM: 609420 MIM: 609422 | ||||||||||||||||||||||||||||||||
Literature Links: |
MIR17 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MIR17 - microRNA 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR17HG - miR-17-92a-1 cluster host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR18A - microRNA 18a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR19A - microRNA 19a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR19B1 - microRNA 19b-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR20A - microRNA 20a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR92A1 - microRNA 92a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |