Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTGGAGTGGGACAAGACTTAGA[A/G]GGTGCAGAAAAGAAAGGGGGCGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603504 | ||||||||||||||||||||
Literature Links: |
CDC14A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC14A - cell division cycle 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319210.1 | Intron | NP_001306139.1 | ||||
NM_001319211.1 | Intron | NP_001306140.1 | ||||
NM_001319212.1 | Intron | NP_001306141.1 | ||||
NM_003672.3 | Intron | NP_003663.2 | ||||
NM_033312.2 | Intron | NP_201569.1 | ||||
NM_033313.2 | Intron | NP_201570.1 | ||||
XM_005271294.3 | Intron | XP_005271351.1 | ||||
XM_005271296.3 | Intron | XP_005271353.1 | ||||
XM_011542340.2 | Intron | XP_011540642.1 | ||||
XM_011542341.2 | Intron | XP_011540643.1 | ||||
XM_011542345.2 | Intron | XP_011540647.1 | ||||
XM_011542346.2 | Intron | XP_011540648.1 | ||||
XM_017002646.1 | Intron | XP_016858135.1 | ||||
XM_017002647.1 | Intron | XP_016858136.1 |