Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTGCGGTGTTGTTAGCAGCACC[A/G]TAATGAAGGCTTTGTGGAGGGATAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608215 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LHX6 - LIM homeobox 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242333.1 | 1567 | Intron | NP_001229262.1 | |||
NM_001242334.1 | 1567 | Intron | NP_001229263.1 | |||
NM_001242335.1 | 1567 | Intron | NP_001229264.1 | |||
NM_014368.4 | 1567 | Intron | NP_055183.2 | |||
NM_199160.3 | 1567 | Intron | NP_954629.2 | |||
XM_005251916.3 | 1567 | Intron | XP_005251973.1 | |||
XM_011518521.2 | 1567 | Silent Mutation | TAC,TAT | Y,Y 382 | XP_011516823.1 | |
XM_011518522.2 | 1567 | Silent Mutation | TAC,TAT | Y,Y 353 | XP_011516824.1 |
MORN5 - MORN repeat containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286828.1 | 1567 | Intron | NP_001273757.1 | |||
NM_198469.3 | 1567 | Intron | NP_940871.2 | |||
XM_005251877.4 | 1567 | Intron | XP_005251934.1 | |||
XM_005251878.3 | 1567 | Intron | XP_005251935.1 | |||
XM_006717034.3 | 1567 | Intron | XP_006717097.1 | |||
XM_011518457.2 | 1567 | Intron | XP_011516759.1 | |||
XM_011518458.1 | 1567 | Intron | XP_011516760.1 | |||
XM_017014578.1 | 1567 | Intron | XP_016870067.1 |
Set Membership: |
HapMap |