Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTAAAGTAGTTTTAGTTGAAGCCT[A/G]CATCAGAATTCTTCAGTGTTTCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604222 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR3 - ARP3 actin related protein 3 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277140.1 | Intron | NP_001264069.1 | ||||
NM_005721.4 | Intron | NP_005712.1 |
LOC101060091 - uncharacterized LOC101060091 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |