Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCTACATCTCCAACACACATT[C/T]GAGAACTACGATAAAAAGCAAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf135 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C9orf135 - chromosome 9 open reading frame 135 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010940.2 | 99 | Intron | NP_001010940.1 | |||
NM_001308084.1 | 99 | Intron | NP_001295013.1 | |||
NM_001308085.1 | 99 | Intron | NP_001295014.1 | |||
NM_001308086.1 | 99 | Intron | NP_001295015.1 | |||
XM_011518229.2 | 99 | Intron | XP_011516531.1 | |||
XM_011518230.2 | 99 | Intron | XP_011516532.1 | |||
XM_011518231.2 | 99 | Intron | XP_011516533.1 | |||
XM_011518233.2 | 99 | UTR 5 | XP_011516535.1 | |||
XM_011518234.2 | 99 | Intron | XP_011516536.1 |
C9orf135-AS1 - C9orf135 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |