Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC6L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC6L2 - ERCC excision repair 6 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010895.2 | 389 | Missense Mutation | ATG,GTG | M,V 1 | NP_001010895.1 | |
NM_020207.4 | 389 | Missense Mutation | ATG,GTG | M,V 1 | NP_064592.2 | |
XM_011518641.2 | 389 | Missense Mutation | ATG,GTG | M,V 1 | XP_011516943.1 | |
XM_011518644.2 | 389 | UTR 5 | XP_011516946.1 | |||
XM_011518645.2 | 389 | Missense Mutation | ATG,GTG | M,V 1 | XP_011516947.1 | |
XM_011518646.2 | 389 | Missense Mutation | ATG,GTG | M,V 1 | XP_011516948.1 | |
XM_011518647.1 | 389 | Missense Mutation | ATG,GTG | M,V 1 | XP_011516949.1 | |
XM_011518648.2 | 389 | Intron | XP_011516950.1 | |||
XM_011518650.2 | 389 | Intron | XP_011516952.1 | |||
XM_017014707.1 | 389 | UTR 5 | XP_016870196.1 |
LINC00476 - long intergenic non-protein coding RNA 476 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |