Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602658 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01537 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LINC01537 - long intergenic non-protein coding RNA 1537 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE2A - phosphodiesterase 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143839.3 | 3080 | Silent Mutation | GCA,GCG | A,A 860 | NP_001137311.1 | |
NM_001146209.2 | 3080 | Silent Mutation | GCA,GCG | A,A 858 | NP_001139681.1 | |
NM_001243784.1 | 3080 | Silent Mutation | GCA,GCG | A,A 846 | NP_001230713.1 | |
NM_002599.4 | 3080 | Silent Mutation | GCA,GCG | A,A 867 | NP_002590.1 | |
XM_005274040.2 | 3080 | Silent Mutation | GCA,GCG | A,A 866 | XP_005274097.1 | |
XM_011545096.2 | 3080 | Silent Mutation | GCA,GCG | A,A 546 | XP_011543398.1 | |
XM_017017910.1 | 3080 | Silent Mutation | GCA,GCG | A,A 546 | XP_016873399.1 |
Set Membership: |
HapMap Validated |