Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602658 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01537 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC01537 - long intergenic non-protein coding RNA 1537 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE2A - phosphodiesterase 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143839.3 | 3134 | Silent Mutation | TAC,TAT | Y,Y 878 | NP_001137311.1 | |
NM_001146209.2 | 3134 | Silent Mutation | TAC,TAT | Y,Y 876 | NP_001139681.1 | |
NM_001243784.1 | 3134 | Silent Mutation | TAC,TAT | Y,Y 864 | NP_001230713.1 | |
NM_002599.4 | 3134 | Silent Mutation | TAC,TAT | Y,Y 885 | NP_002590.1 | |
XM_005274040.2 | 3134 | Nonsense Mutation | XP_005274097.1 | |||
XM_011545096.2 | 3134 | Silent Mutation | TAC,TAT | Y,Y 564 | XP_011543398.1 | |
XM_017017910.1 | 3134 | Silent Mutation | TAC,TAT | Y,Y 564 | XP_016873399.1 |
Set Membership: |
HapMap |