Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGATTTCACTGTTCTTGTGGAG[C/T]TTACATTCACTTTAGTGGAGAAGAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611122 | |||||||||||||||||||||||
Literature Links: |
ANKRD28 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ANKRD28 - ankyrin repeat domain 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195098.1 | Intron | NP_001182027.1 | ||||
NM_001195099.1 | Intron | NP_001182028.1 | ||||
NM_015199.3 | Intron | NP_056014.2 | ||||
XM_005264996.3 | Intron | XP_005265053.1 | ||||
XM_011533540.2 | Intron | XP_011531842.1 | ||||
XM_011533541.2 | Intron | XP_011531843.1 | ||||
XM_011533542.2 | Intron | XP_011531844.1 | ||||
XM_011533543.2 | Intron | XP_011531845.1 | ||||
XM_011533545.2 | Intron | XP_011531847.1 | ||||
XM_011533546.2 | Intron | XP_011531848.1 | ||||
XM_011533547.2 | Intron | XP_011531849.1 | ||||
XM_017006026.1 | Intron | XP_016861515.1 | ||||
XM_017006027.1 | Intron | XP_016861516.1 | ||||
XM_017006028.1 | Intron | XP_016861517.1 | ||||
XM_017006029.1 | Intron | XP_016861518.1 |