Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCTCCACTGTTTACTGAGTTA[C/T]CTGGGGAAAGTTACCTAATTTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608080 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MRO - maestro | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127174.1 | 1638 | UTR 3 | NP_001120646.1 | |||
NM_001127175.1 | 1638 | UTR 3 | NP_001120647.1 | |||
NM_001127176.1 | 1638 | UTR 3 | NP_001120648.1 | |||
NM_031939.3 | 1638 | UTR 3 | NP_114145.2 | |||
XM_005258365.3 | 1638 | UTR 3 | XP_005258422.1 | |||
XM_011526226.2 | 1638 | UTR 3 | XP_011524528.1 | |||
XM_017026035.1 | 1638 | UTR 3 | XP_016881524.1 | |||
XM_017026036.1 | 1638 | UTR 3 | XP_016881525.1 | |||
XM_017026037.1 | 1638 | UTR 3 | XP_016881526.1 |