Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTATGAAGACAGTCTATTCTCT[A/G]TTTTGTTCCTTCCTCTAGGGAGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAA40 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAA40 - N(alpha)-acetyltransferase 40, NatD catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300800.1 | Intron | NP_001287729.1 | ||||
NM_024771.3 | Intron | NP_079047.2 | ||||
XM_006718689.3 | Intron | XP_006718752.1 | ||||
XM_011545254.2 | Intron | XP_011543556.1 | ||||
XM_011545255.2 | Intron | XP_011543557.1 |