Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610400 MIM: 608337 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MACROD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MACROD1 - MACRO domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014067.3 | Intron | NP_054786.2 | ||||
XM_005273939.4 | Intron | XP_005273996.1 | ||||
XM_005273940.2 | Intron | XP_005273997.1 | ||||
XM_006718521.3 | Intron | XP_006718584.1 | ||||
XM_006718522.2 | Intron | XP_006718585.1 | ||||
XM_011544970.2 | Intron | XP_011543272.1 |
OTUB1 - OTU deubiquitinase, ubiquitin aldehyde binding 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |