Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTATTTCACAAAAGGTGGTGGGA[A/G]AGAACTCAAAAACGCTGACACCTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616381 | ||||||||||||||||||||
Literature Links: |
RSRC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSRC2 - arginine and serine rich coiled-coil 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023012.5 | Intron | NP_075388.2 | ||||
XM_005253601.1 | Intron | XP_005253658.1 | ||||
XM_005253602.2 | Intron | XP_005253659.1 | ||||
XM_005253604.1 | Intron | XP_005253661.1 | ||||
XM_011538688.1 | Intron | XP_011536990.1 | ||||
XM_011538689.1 | Intron | XP_011536991.1 | ||||
XM_017019830.1 | Intron | XP_016875319.1 | ||||
XM_017019831.1 | Intron | XP_016875320.1 | ||||
XM_017019832.1 | Intron | XP_016875321.1 | ||||
XM_017019833.1 | Intron | XP_016875322.1 |
ZCCHC8 - zinc finger CCHC-type containing 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |