Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCTTGGTGGAGCCATAGGCCGC[A/G]TGCACGCGCAGGTGCAGCTGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607668 MIM: 608920 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL6IP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARL6IP4 - ADP ribosylation factor like GTPase 6 interacting protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OGFOD2 - 2-oxoglutarate and iron dependent oxygenase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNM2 - phosphatidylinositol transfer protein membrane associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300801.1 | 4076 | Silent Mutation | CAC,CAT | H,H 1199 | NP_001287730.1 | |
NM_020845.2 | 4076 | Silent Mutation | CAC,CAT | H,H 1205 | NP_065896.1 | |
XM_011538590.2 | 4076 | Silent Mutation | CAC,CAT | H,H 1238 | XP_011536892.1 |
Set Membership: |
Validated |