Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGGGGAGCAGGTAGCAAGTGTA[G/T]ACCGAGAACATTAAAAGGAGATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601950 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFPM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFPM1 - zinc finger protein, FOG family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153813.2 | Intron | NP_722520.2 | ||||
XM_011522912.2 | Intron | XP_011521214.1 | ||||
XM_011522914.2 | Intron | XP_011521216.1 | ||||
XM_011522917.2 | Intron | XP_011521219.1 | ||||
XM_017022982.1 | Intron | XP_016878471.1 |
Set Membership: |
HapMap |