Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611675 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0513 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA0513 - KIAA0513 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286565.1 | Intron | NP_001273494.1 | ||||
NM_001286566.1 | Intron | NP_001273495.1 | ||||
NM_001297766.1 | Intron | NP_001284695.1 | ||||
NM_014732.3 | Intron | NP_055547.1 | ||||
XM_005256265.3 | Intron | XP_005256322.1 | ||||
XM_017023912.1 | Intron | XP_016879401.1 |