Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGGATGGCCTGCCTCCTTCGC[G/T]CATTTCAGAGAATTTCTGCAGGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606435 MIM: 606431 MIM: 606432 MIM: 606433 MIM: 606434 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT1A10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
UGT1A6*2a,c.19T>G UGT1A6*2a,g.19T>G UGT1A6*2b,c.19T>G UGT1A6*2b,g.19T>G UGT1A6*2c,c.19T>G UGT1A6*2c,g.19T>G UGT1A6*2d,c.19T>G UGT1A6*2d,g.19T>G UGT1A6*2e,c.19T>G UGT1A6*2e,g.19T>G UGT1A6*3a,c.19T>G UGT1A6*3a,g.19T>G UGT1A6*3b,c.19T>G UGT1A6*3b,g.19T>G UGT1A6*4,c.19T>G UGT1A6*4a,c.19T>G UGT1A6*4a,g.19T>G UGT1A6*4b,c.19T>G UGT1A6*4b,g.19T>G UGT1A6*4c,c.19T>G UGT1A6*4c,g.19T>G UGT1A6*5,c.19T>G UGT1A6*6,c.19T>G UGT1A6*6,g.19T>G UGT1A6*7,c.19T>G UGT1A6*7,g.19T>G | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
UGT1A10 - UDP glucuronosyltransferase family 1 member A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019075.2 | 158 | Intron | NP_061948.1 |
UGT1A6 - UDP glucuronosyltransferase family 1 member A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001072.3 | 158 | Missense Mutation | GCA,TCA | A,S 7 | NP_001063.2 | |
NM_205862.1 | 158 | Intron | NP_995584.1 |
UGT1A7 - UDP glucuronosyltransferase family 1 member A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019077.2 | 158 | Intron | NP_061950.2 |
UGT1A8 - UDP glucuronosyltransferase family 1 member A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019076.4 | 158 | Intron | NP_061949.3 |
UGT1A9 - UDP glucuronosyltransferase family 1 member A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021027.2 | 158 | Intron | NP_066307.1 |
Set Membership: |
HapMap DME Validated Inventoried |