Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605173 MIM: 606873 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ENC1 - ectodermal-neural cortex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256574.1 | Intron | NP_001243503.1 | ||||
NM_001256575.1 | Intron | NP_001243504.1 | ||||
NM_001256576.1 | Intron | NP_001243505.1 | ||||
NM_003633.3 | Intron | NP_003624.1 | ||||
XM_011543696.2 | Intron | XP_011541998.1 | ||||
XM_011543697.2 | Intron | XP_011541999.1 |
HEXB - hexosaminidase subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000521.3 | Intron | NP_000512.1 | ||||
NM_001292004.1 | Intron | NP_001278933.1 |
Set Membership: |
HapMap Validated |