Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605173 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ENC1 - ectodermal-neural cortex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256574.1 | 4900 | UTR 3 | NP_001243503.1 | |||
NM_001256575.1 | 4900 | UTR 3 | NP_001243504.1 | |||
NM_001256576.1 | 4900 | UTR 3 | NP_001243505.1 | |||
NM_003633.3 | 4900 | UTR 3 | NP_003624.1 | |||
XM_011543696.2 | 4900 | UTR 3 | XP_011541998.1 | |||
XM_011543697.2 | 4900 | UTR 3 | XP_011541999.1 |