Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGAACGTAGCCCTGCTTAGCC[C/T]TGCTGTCGGTTGTGGGAGGCTCTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300762 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEB16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAGEB16 - MAGE family member B16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099921.1 | Intron | NP_001093391.1 | ||||
XM_011545453.2 | Intron | XP_011543755.1 | ||||
XM_011545454.2 | Intron | XP_011543756.1 |
Set Membership: |
HapMap |