Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGAGTAAGTTAGAGGTCAGCA[A/G]TGAGGGCTGGCCTGTGCAGGTGTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120130 MIM: 120090 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
COL4A1 - collagen type IV alpha 1 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COL4A2 - collagen type IV alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |