Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133510 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ERCC3 - ERCC excision repair 3, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000122.1 | Intron | NP_000113.1 | ||||
NM_001303416.1 | Intron | NP_001290345.1 | ||||
NM_001303418.1 | Intron | NP_001290347.1 | ||||
XM_011510794.1 | Intron | XP_011509096.1 | ||||
XM_011510795.1 | Intron | XP_011509097.1 | ||||
XM_017003583.1 | Intron | XP_016859072.1 |
Set Membership: |
HapMap Validated |