Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTGGGCTCAGAAGGAGCCCAC[A/G]GATTTAATCTTCTTGTTAAGAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605684 MIM: 608487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM34 - tripartite motif containing 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003827.1 | Intron | NP_001003827.1 | ||||
NM_021616.5 | Intron | NP_067629.2 | ||||
NM_130390.2 | Intron | NP_569074.2 |
TRIM5 - tripartite motif containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033034.2 | Intron | NP_149023.2 | ||||
NM_033092.2 | Intron | NP_149083.2 | ||||
NM_033093.2 | Intron | NP_149084.2 | ||||
XM_005253183.2 | Intron | XP_005253240.1 | ||||
XM_005253184.2 | Intron | XP_005253241.1 | ||||
XM_006718358.2 | Intron | XP_006718421.1 | ||||
XM_011520426.2 | Intron | XP_011518728.1 | ||||
XM_011520427.1 | Intron | XP_011518729.1 | ||||
XM_017018460.1 | Intron | XP_016873949.1 | ||||
XM_017018461.1 | Intron | XP_016873950.1 | ||||
XM_017018462.1 | Intron | XP_016873951.1 | ||||
XM_017018463.1 | Intron | XP_016873952.1 |
TRIM6-TRIM34 - TRIM6-TRIM34 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003819.3 | Intron | NP_001003819.1 |