Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606518 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAVCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HAVCR1 - hepatitis A virus cellular receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173393.2 | Intron | NP_001166864.1 | ||||
NM_001308156.1 | Intron | NP_001295085.1 | ||||
NM_012206.3 | Intron | NP_036338.2 | ||||
XM_011534515.2 | Intron | XP_011532817.1 | ||||
XM_017009339.1 | Intron | XP_016864828.1 | ||||
XM_017009340.1 | Intron | XP_016864829.1 |