Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGAGCCATTTGTAACCTTAAAT[C/G]GTTCTTTTAGAGAACAAAGACTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF891 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF891 - zinc finger protein 891 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277291.1 | 1911 | UTR 3 | NP_001264220.1 | |||
XM_017018666.1 | 1911 | UTR 3 | XP_016874155.1 | |||
XM_017018667.1 | 1911 | UTR 3 | XP_016874156.1 | |||
XM_017018668.1 | 1911 | UTR 3 | XP_016874157.1 |
Set Membership: |
HapMap |