Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603251 MIM: 613405 MIM: 604722 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CDK9 - cyclin dependent kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261.3 | 267 | Intron | NP_001252.1 | |||
XM_017014184.1 | 267 | UTR 5 | XP_016869673.1 |
MIR2861 - microRNA 2861 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3960 - microRNA 3960 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2D3C - SH2 domain containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |