Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGCCCTGCCTGCCCAGCACGG[A/C]AAAAAATCACTCTTCTGGCCCCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612034 MIM: 180535 | ||||||||||||||||||||
Literature Links: |
APC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APC2 - APC2, WNT signaling pathway regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005883.2 | Intron | NP_005874.1 | ||||
XM_005259475.2 | Intron | XP_005259532.1 | ||||
XM_006722607.2 | Intron | XP_006722670.1 | ||||
XM_006722608.3 | Intron | XP_006722671.2 | ||||
XM_006722609.3 | Intron | XP_006722672.1 | ||||
XM_006722610.3 | Intron | XP_006722673.2 |
RPS15 - ribosomal protein S15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |