Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGAGGCCAAGGCTTGAGATAGA[G/T]TGGAGGGGGCCGGAGAATACTAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602702 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLIN3 - perilipin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164189.1 | Intron | NP_001157661.1 | ||||
NM_001164194.1 | Intron | NP_001157666.1 | ||||
NM_005817.4 | Intron | NP_005808.3 |
Set Membership: |
HapMap |