Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC1A1 - solute carrier family 1 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004170.5 | Intron | NP_004161.4 | ||||
XM_011518007.1 | Intron | XP_011516309.1 | ||||
XM_011518008.2 | Intron | XP_011516310.1 | ||||
XM_011518009.2 | Intron | XP_011516311.1 | ||||
XM_011518010.1 | Intron | XP_011516312.1 | ||||
XM_017015042.1 | Intron | XP_016870531.1 | ||||
XM_017015043.1 | Intron | XP_016870532.1 |
Set Membership: |
HapMap |