Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAAAATGCAGTCTCCCATCCAG[A/G]TCCTCAGACCCACACACCAGCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNN3 - potassium calcium-activated channel subfamily N member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204087.1 | 12826 | UTR 3 | NP_001191016.1 | |||
NM_002249.5 | 12826 | UTR 3 | NP_002240.3 | |||
NM_170782.2 | 12826 | UTR 3 | NP_740752.1 |