Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604884 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC613206 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC613206 - myeloproliferative disease associated tumor antigen 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEK6 - NIMA related kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145001.2 | Intron | NP_001138473.1 | ||||
NM_001166167.1 | Intron | NP_001159639.1 | ||||
NM_001166168.1 | Intron | NP_001159640.1 | ||||
NM_001166169.1 | Intron | NP_001159641.1 | ||||
NM_001166170.1 | Intron | NP_001159642.1 | ||||
NM_001166171.1 | Intron | NP_001159643.1 | ||||
NM_014397.5 | Intron | NP_055212.2 | ||||
XM_005251664.1 | Intron | XP_005251721.1 | ||||
XM_006716936.2 | Intron | XP_006716999.1 | ||||
XM_017014217.1 | Intron | XP_016869706.1 |
Set Membership: |
HapMap Validated |