Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGCTCCTTATTGCGCAGGCGCAA[A/G]GACTGCCCGGTAGGACGAGCGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600187 MIM: 601935 MIM: 615865 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EIF5A - eukaryotic translation initiation factor 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143760.1 | Intron | NP_001137232.1 | ||||
NM_001143761.1 | Intron | NP_001137233.1 | ||||
NM_001143762.1 | Intron | NP_001137234.1 | ||||
NM_001970.4 | Intron | NP_001961.1 | ||||
XM_005256509.2 | Intron | XP_005256566.1 | ||||
XM_011523710.2 | Intron | XP_011522012.1 | ||||
XM_011523711.2 | Intron | XP_011522013.1 | ||||
XM_011523712.2 | Intron | XP_011522014.1 | ||||
XM_011523713.2 | Intron | XP_011522015.1 | ||||
XM_017024300.1 | Intron | XP_016879789.1 | ||||
XM_017024301.1 | Intron | XP_016879790.1 |
GPS2 - G protein pathway suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEURL4 - neuralized E3 ubiquitin protein ligase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |