Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTGTCTCGGGGAGGCGCGGTT[A/C]GAGCCGGAATCAGCCTCCCTCCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM83B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM83B - family with sequence similarity 83 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010872.2 | 43 | Intron | NP_001010872.1 | |||
XM_006715022.3 | 43 | UTR 5 | XP_006715085.1 | |||
XM_011514394.2 | 43 | Intron | XP_011512696.1 | |||
XM_011514395.2 | 43 | Intron | XP_011512697.1 | |||
XM_017010478.1 | 43 | Intron | XP_016865967.1 |