Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAACAAGCTAATCAACCACCACT[C/G]TTTCTACTATCCATAATAATAAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606066 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LHX9 - LIM homeobox 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014434.1 | Intron | NP_001014434.1 | ||||
NM_020204.2 | Intron | NP_064589.2 | ||||
XM_005245350.3 | Intron | XP_005245407.2 | ||||
XM_011509781.2 | Intron | XP_011508083.2 | ||||
XM_017001849.1 | Intron | XP_016857338.1 |
Set Membership: |
HapMap |