Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603182 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ILF3 - interleukin enhancer binding factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137673.1 | Intron | NP_001131145.1 | ||||
NM_004516.3 | Intron | NP_004507.2 | ||||
NM_012218.3 | Intron | NP_036350.2 | ||||
NM_017620.2 | Intron | NP_060090.2 | ||||
NM_153464.2 | Intron | NP_703194.1 | ||||
XM_006722742.2 | Intron | XP_006722805.1 | ||||
XM_011527984.2 | Intron | XP_011526286.2 | ||||
XM_011527985.2 | Intron | XP_011526287.1 | ||||
XM_011527987.1 | Intron | XP_011526289.1 | ||||
XM_017026763.1 | Intron | XP_016882252.1 | ||||
XM_017026764.1 | Intron | XP_016882253.1 | ||||
XM_017026765.1 | Intron | XP_016882254.1 |
Set Membership: |
HapMap |