Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTGAATTTCAGAGCCTAATTTG[C/G]TCTCTCTCAAGTGAATATCCACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603182 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ILF3 - interleukin enhancer binding factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137673.1 | Intron | NP_001131145.1 | ||||
NM_004516.3 | Intron | NP_004507.2 | ||||
NM_012218.3 | Intron | NP_036350.2 | ||||
NM_017620.2 | Intron | NP_060090.2 | ||||
NM_153464.2 | Intron | NP_703194.1 | ||||
XM_006722742.2 | Intron | XP_006722805.1 | ||||
XM_011527984.2 | Intron | XP_011526286.2 | ||||
XM_011527985.2 | Intron | XP_011526287.1 | ||||
XM_011527987.1 | Intron | XP_011526289.1 | ||||
XM_017026763.1 | Intron | XP_016882252.1 | ||||
XM_017026764.1 | Intron | XP_016882253.1 | ||||
XM_017026765.1 | Intron | XP_016882254.1 |
ILF3-AS1 - ILF3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |