Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTGGGCACTTATTCACACTTA[C/T]AATACTTATGGCTTCATACAGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612134 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLCE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLCE - glucuronic acid epimerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324091.1 | Intron | NP_001311020.1 | ||||
NM_001324092.1 | Intron | NP_001311021.1 | ||||
NM_001324093.1 | Intron | NP_001311022.1 | ||||
NM_001324094.1 | Intron | NP_001311023.1 | ||||
NM_015554.2 | Intron | NP_056369.1 | ||||
XM_005254298.3 | Intron | XP_005254355.1 | ||||
XM_017022073.1 | Intron | XP_016877562.1 | ||||
XM_017022074.1 | Intron | XP_016877563.1 |
Set Membership: |
HapMap |