Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611104 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGD4 - FYVE, RhoGEF and PH domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304480.1 | 43 | Intron | NP_001291409.1 | |||
NM_001304481.1 | 43 | Intron | NP_001291410.1 | |||
NM_001304483.1 | 43 | Intron | NP_001291412.1 | |||
NM_001304484.1 | 43 | Intron | NP_001291413.1 | |||
NM_139241.3 | 43 | Intron | NP_640334.2 | |||
XM_005253304.4 | 43 | UTR 5 | XP_005253361.1 | |||
XM_005253307.3 | 43 | Intron | XP_005253364.1 | |||
XM_005253308.4 | 43 | Intron | XP_005253365.1 | |||
XM_005253309.1 | 43 | Intron | XP_005253366.1 | |||
XM_005253310.3 | 43 | Intron | XP_005253367.1 | |||
XM_011520554.1 | 43 | Intron | XP_011518856.1 | |||
XM_011520555.1 | 43 | Intron | XP_011518857.1 | |||
XM_011520556.1 | 43 | Intron | XP_011518858.1 | |||
XM_011520557.1 | 43 | Intron | XP_011518859.1 | |||
XM_011520558.2 | 43 | Intron | XP_011518860.1 | |||
XM_011520559.2 | 43 | Intron | XP_011518861.1 | |||
XM_017018803.1 | 43 | UTR 5 | XP_016874292.1 | |||
XM_017018804.1 | 43 | Intron | XP_016874293.1 | |||
XM_017018805.1 | 43 | Intron | XP_016874294.1 |