Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609594 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VEPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VEPH1 - ventricular zone expressed PH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167911.1 | Intron | NP_001161383.1 | ||||
NM_001167912.1 | Intron | NP_001161384.1 | ||||
NM_001167915.1 | Intron | NP_001161387.1 | ||||
NM_001167916.1 | Intron | NP_001161388.1 | ||||
NM_001167917.1 | Intron | NP_001161389.1 | ||||
NM_024621.2 | Intron | NP_078897.2 | ||||
XM_011513134.2 | Intron | XP_011511436.1 | ||||
XM_011513135.1 | Intron | XP_011511437.1 |