Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCAGACATGCCTTCAGTCAGCCC[G/T]GGTCCCCAGCGGCCTCCCAAGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603809 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3D21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH3D21 - SH3 domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162530.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | NP_001156002.1 | |
NM_024676.4 | 544 | Silent Mutation | CCG,CCT | P,P 46 | NP_078952.4 | |
XM_011542153.2 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_011540455.1 | |
XM_017002340.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_016857829.1 | |
XM_017002341.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_016857830.1 | |
XM_017002342.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_016857831.1 | |
XM_017002343.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_016857832.1 | |
XM_017002344.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_016857833.1 | |
XM_017002345.1 | 544 | Silent Mutation | CCG,CCT | P,P 157 | XP_016857834.1 | |
XM_017002346.1 | 544 | UTR 5 | XP_016857835.1 |
THRAP3 - thyroid hormone receptor associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |