Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZCCHC17 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZCCHC17 - zinc finger CCHC-type containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282566.1 | Intron | NP_001269495.1 | ||||
NM_001282567.1 | Intron | NP_001269496.1 | ||||
NM_001282568.1 | Intron | NP_001269497.1 | ||||
NM_001282569.1 | Intron | NP_001269498.1 | ||||
NM_001282570.1 | Intron | NP_001269499.1 | ||||
NM_001282571.1 | Intron | NP_001269500.1 | ||||
NM_001282572.1 | Intron | NP_001269501.1 | ||||
NM_001282573.1 | Intron | NP_001269502.1 | ||||
NM_001282574.1 | Intron | NP_001269503.1 | ||||
NM_016505.3 | Intron | NP_057589.2 | ||||
XM_006710681.3 | Intron | XP_006710744.1 | ||||
XM_011541567.2 | Intron | XP_011539869.1 |